• USA Home
  • 446343 - Methylbis(trimethylsilyloxy)vinylsilane

EMAIL THIS PAGE TO A FRIEND
446343 Aldrich

Methylbis(trimethylsilyloxy)vinylsilane

98%

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   MTNTUAHMWISEEF-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.397(lit.)
bp   48 °C/8.8 mmHg(lit.)
density   0.864 g/mL at 25 °C(lit.)

Description

Packaging

25 mL in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
123.8 °F
Flash Point(C) 
51 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?