EMAIL THIS PAGE TO A FRIEND
446475 Aldrich

Ajmalicine

97%

Synonym: δ-Yohimbine, Py-Tetrahydroserpentine, Raubasine

  • CAS Number 483-04-5

  • Empirical Formula (Hill Notation) C21H24N2O3

  • Molecular Weight 352.43

  •  Beilstein Registry Number 97268

  •  EC Number 207-589-5

  •  PubChem Substance ID 329756528

Purchase

Properties

InChI Key   GRTOGORTSDXSFK-QKEZJILASA-N
assay   97%

Description

Biochem/physiol Actions

α1 adrenoceptor antagonist.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
AX7875000

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Yohimbe Alkaloids on Ascentis® Express F5, HILIC Mode

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Yohimbe Alkaloids on Ascentis® Express F5, HILIC Mode
Keywords: Chromatography, High performance liquid chromatography, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?