• USA Home
  • 446548 - Dow Corning® silicone diffusion pump fluids

EMAIL THIS PAGE TO A FRIEND
446548 Aldrich

Dow Corning® silicone diffusion pump fluids

702

  • Linear Formula [CH3Si(C6H5)2O]2Si(CH3)2

  •  MDL number MFCD00053661

Purchase

Properties

Related Categories Aldrich swivel McLeod gauge, Chemistry Equipment and Labware, Labware, Vacuum Equipment, Vacuum Gauges and Manometers More...
InChI Key   YFCVAZGXPLMNDG-UHFFFAOYSA-N
refractive index   n20/D 1.52
bp   180 °C/0.5 mmHg
density   1.07 g/mL at 25 °C

Description

Application

General purpose for pumping large volumes of gas, vacuum range 10-5 to 10-7 torr. Also suitable for use in Aldrich swivel McLeod vacuum gauges.

General description

For high vacuum and for fast pumping of large volumes of gas or vapor in production operations. Chemically and thermally inert fluids. General purpose for pumping large volumes of gas, vacuum range 10-5 to 10-7 torr. Also suitable for use in Aldrich swivel McLeod vacuum gauges.

Legal Information

Dow Corning is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?