Synonym: EMIMBF4
CAS Number 143314-16-3
Empirical Formula (Hill Notation) C6H11BF4N2
Molecular Weight 197.97
MDL number MFCD00216668
PubChem Substance ID 24868356
Popular Documents: FTIR (PDF)
| Related Categories | Chemical Synthesis, Imidazolium, Ionic Liquids, Specialty Synthesis |
| assay | 97% |
| refractive index |
n |
| bp | >350 °C(lit.) |
| mp | 15 °C(lit.) |
| density | 1.294 g/mL at 25 °C(lit.) |
5 mL in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?