EMAIL THIS PAGE TO A FRIEND
449970 Aldrich

Potassium bromide

anhydrous, beads, −10 mesh, ≥99.9% trace metals basis

Purchase

Properties

Related Categories 19: K, Beaded Materials, Chemical Synthesis, Crystal Grade Inorganics, Essential Chemicals,
grade   anhydrous
vapor pressure   <0.01 mmHg ( 20 °C)
  1 mmHg ( 795 °C)
  1.853 Pa ( 20 °C)(lit.)
InChI Key   IOLCXVTUBQKXJR-UHFFFAOYSA-M
assay   ≥99.9% trace metals basis
form   beads
refractive index   n/D 1.5598 (at 589nm)(lit.)
particle size   −10 mesh
bp   1435 °C/1 atm(lit.)
mp   734 °C(lit.)
solubility   ethanol: slightly soluble(lit.)

Description

Application

May be used for removal of peripheral membrane proteins.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TS7650000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

related product

Product #

Description

Add to Cart

94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?