EMAIL THIS PAGE TO A FRIEND
451096 Aldrich

Potassium

rod, diam. 25 mm, 99.5%, in mineral oil

  • CAS Number 7440-09-7

  • Empirical Formula (Hill Notation) K

  • Molecular Weight 39.10

  •  MDL number MFCD00133776

Purchase

Properties

vapor pressure   0.09 mmHg ( 260 °C)
InChI Key   ZLMJMSJWJFRBEC-UHFFFAOYSA-N
assay   99.5%
form   rod
resistivity   6.1 μΩ-cm, 20°C
diam.   25 mm
bp   760 °C(lit.)
mp   64 °C(lit.)
density   0.86 g/mL at 25 °C(lit.)

Description

Application

Combines under pressure with silver metal to form 2 new compounds: K2Ag and K3Ag.†

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 2257 4.3 / PGI
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

679909 Potassium, cubes (in mineral oil), L × W × H 40 mm × 30 mm × 20 mm, 99.5% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?