• USA Home
  • 452076 - cis-2,3-Diphenyl-1-propylaziridine

EMAIL THIS PAGE TO A FRIEND
452076 Aldrich

cis-2,3-Diphenyl-1-propylaziridine

97%

Purchase

Properties

Related Categories Aziridines, Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   BLOUNKOJLURQAH-JWTNVVGKSA-N
assay   97%
contains   1-3% sodium hydroxide as inhibitor
mp   55-57 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?