• USA Home
  • 452092 - 5-Formyl-2-furansulfonic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
452092 Aldrich

5-Formyl-2-furansulfonic acid sodium salt

98%

Synonym: Furfural-5-sulfonic acid sodium salt

Purchase

Properties

InChI Key   ZTJAVDOZUQPVAV-UHFFFAOYSA-M
assay   98%
mp   >300 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?