• USA Home
  • 452238 - Chloro[2-(3-cyclohexen-1-yl)ethyl]dimethylsilane

EMAIL THIS PAGE TO A FRIEND
452238 Aldrich

Chloro[2-(3-cyclohexen-1-yl)ethyl]dimethylsilane

technical grade, 90%

Purchase

Properties

Related Categories Chemical Synthesis, Protecting and Derivatizing Reagents, Protection and Derivatization, Silicon-Based, Synthetic Reagents More...
grade   technical grade
InChI Key   XQRFNPGYTWWODZ-UHFFFAOYSA-N
assay   90%
refractive index   n20/D 1.474(lit.)
bp   119-120 °C/21 mmHg(lit.)
density   0.956 g/mL at 25 °C(lit.)

Description

Packaging

100 mL in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 2987 8 / PGII
WGK Germany 
3
Flash Point(F) 
204.8 °F
Flash Point(C) 
96 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?