EMAIL THIS PAGE TO A FRIEND
452696 Aldrich

p-Carborane

98%

Synonym: 1,12-Dicarbadodecaborane(12)

  • CAS Number 20644-12-6

  • Empirical Formula (Hill Notation) C2H12B10

  • Molecular Weight 144.23

  •  MDL number MFCD00274275

Purchase

Properties

assay   98%
mp   200-203 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?