EMAIL THIS PAGE TO A FRIEND
453331 Aldrich

Deuterium oxide

100.0 atom % D, ≥99.96 atom % D

Synonym: Heavy water, Water-d2

Purchase

Properties

InChI Key   XLYOFNOQVPJJNP-ZSJDYOACSA-N
isotopic purity   ≥99.96 atom % D
  100.0 atom % D
packaging   pk of 10 × 0.25 mL
refractive index   n20/D 1.328(lit.)
bp   101.4 °C(lit.)
mp   3.8 °C(lit.)
density   1.107 g/mL at 25 °C(lit.)

Description

Packaging

Packaged in prescored ampules

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
ZC0230000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules

replaced by

Product #

Description

Add to Cart

269786 Deuterium oxide, "100%", 99.96 atom % D

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?