• USA Home
  • 453412 - (1R,2S,4R)-(−)-4-(1-Methylethyl)-2-(1-methyl-1-phenylethyl)cyclohexanol

EMAIL THIS PAGE TO A FRIEND
453412 Aldrich

(1R,2S,4R)-(−)-4-(1-Methylethyl)-2-(1-methyl-1-phenylethyl)cyclohexanol

98%

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   ZJHNUUVRRFRSFM-DJIMGWMZSA-N
assay   98%
optical activity   [α]20/D −35°, c = 1 in ethanol
optical purity   ee: 99% (HPLC)
mp   68-71 °C(lit.)

Description

Packaging

1 g in glass bottle

Application

Cyclohexyl-based chiral auxiliary that provides equal or greater levels of asymmetric induction when compared to menthol or 8-phenylmenthol in a variety of reactions.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?