• USA Home
  • 454087 - (Methoxymethyl)diphenylphosphine oxide

EMAIL THIS PAGE TO A FRIEND
454087 Aldrich

(Methoxymethyl)diphenylphosphine oxide

98%

Synonym: Diphenyl(methoxymethyl)phosphine oxide

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Phosphine Ligands
InChI Key   OEPKDBQOTLDTNC-UHFFFAOYSA-N
assay   98%
mp   115-117 °C(lit.)

Description

Packaging

10 g in glass bottle

Application

Catalyst for:
• Molybdenum hexacarbonyl-mediated Pauson-Khand reactions
• Rhodium-catalyzed hydroformylation reaction

Price and Availability


Laboratory Gloves

Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
SZ0199360
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?