• USA Home
  • 455458 - Ammonium tetrakis(4-methoxyphenyl)borate hydrate

EMAIL THIS PAGE TO A FRIEND
455458 Aldrich

Ammonium tetrakis(4-methoxyphenyl)borate hydrate

  • CAS Number 314075-09-7

  • Linear Formula NH4B(C6H4OCH3)4 · xH2O

  • Molecular Weight 457.37 (anhydrous basis)

  •  MDL number MFCD01321234

  •  PubChem Substance ID 24869071

Purchase

Properties

Related Categories Boron, Catalysis and Inorganic Chemistry, Chemical Synthesis More...
InChI Key   PACUGZVYJCUKSM-UHFFFAOYSA-O
mp   198 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?