EMAIL THIS PAGE TO A FRIEND
455911 Aldrich

Copper(I) cyanide

99.98%

Synonym: Cuprous cyanide

Purchase

Properties

Related Categories Copper Salts, Essential Chemicals, Materials Science, Metal and Ceramic Science, Research Essentials,
InChI Key   DULSAGLWMRMKCQ-UHFFFAOYSA-N
assay   99.98%
mp   474 °C(lit.)
density   2.92 g/mL at 25 °C(lit.)

Description

Application

Applied in a widely useful, thiosulfate-assisted, synthesis of a variety of diamine-CuCN complexes.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
RIDADR 
UN 1587 6.1 / PGII
WGK Germany 
2
Protocols & Articles

Articles

Reagents for C–C Bond Formation

Aldrich carries a large variety of electrophiles and nucleophiles that are widely used in C–C bond-forming reactions. This group of products contains many organometallic reagents as well as commonly-...
ChemFiles Volume 1 Article 3
Keywords: Acylations, Alkylations

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?