• USA Home
  • 456551 - Poly(1,3-propylene succinate)

EMAIL THIS PAGE TO A FRIEND
456551 Aldrich

Poly(1,3-propylene succinate)

average Mw ~9,500 by GPC

Purchase

Properties

Related Categories Esters, Hydrophobic Polymers, Materials Science, Polymer Science, Polymers More...
InChI Key   OXSQDJLFMPMKNO-UHFFFAOYSA-N
mol wt   average Mw ~9,500 by GPC
transition temp   Tm 51 °C (DSC at peak)

Description

Packaging

10 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?