• USA Home
  • 458082 - Poly(propylene glycol) dibenzoate

EMAIL THIS PAGE TO A FRIEND
458082 Aldrich

Poly(propylene glycol) dibenzoate

average Mn ~400

Purchase

Properties

Related Categories Materials Science, Plasticizers, Polymer Additives, Polymer Science
mol wt   average Mn ~400
refractive index   n20/D 1.518(lit.)
bp   263 °C/5 mmHg(lit.)
density   1.103 g/mL at 25 °C(lit.)

Description

Application

Plasticizer

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?