• USA Home
  • 458155 - Poly[ethyl acrylate-co-methacrylic acid-co-3-(1-isocyanato-1-methylethyl)-α-methylstyrene], adduct with ethoxylated nonylphenol solution

EMAIL THIS PAGE TO A FRIEND
458155 Aldrich

Poly[ethyl acrylate-co-methacrylic acid-co-3-(1-isocyanato-1-methylethyl)-α-methylstyrene], adduct with ethoxylated nonylphenol solution

25 wt. % in H2O

Purchase

Properties

Related Categories Hydrophilic Polymers, Materials Science, Poly(acrylic acid), Polymethacrylate and Other Acrylic Polymers, Poly(methacrylic acid) (PMAA) Copolymers, Poly(methacrylic acid)s and Polymethacrylates,
concentration   25 wt. % in H2O
pH   2.5-4.5
useful pH range   7 - 11
viscosity   10-100 cP(lit.)
bp   73 °C
density   1.1 g/mL at 25 °C

Description

Application

Alkali soluble associative thickener.
Thickener for aqueous-based products.

Features and Benefits

Hydrophobe modified ethoxylate urethane-alkali soluble emulsion

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?