EMAIL THIS PAGE TO A FRIEND
459062 Aldrich

Tungstic acid

99.999% trace metals basis

Purchase

Properties

Related Categories Acids, Electronic Chemicals, Essential Chemicals, Materials Science, Metal and Ceramic Science,
InChI Key   CMPGARWFYBADJI-UHFFFAOYSA-L
assay   99.999% trace metals basis
form   powder
density   5.5 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

223328 Tungstic acid, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?