• USA Home
  • 459127 - 2-(Trifluoromethyl)phenylhydrazine

EMAIL THIS PAGE TO A FRIEND
459127 Aldrich

2-(Trifluoromethyl)phenylhydrazine

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Hydrazines, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   JSWQDLBFVSTSIW-UHFFFAOYSA-N
assay   97%
mp   56-60 °C(lit.)

Description

Packaging

5 g in glass bottle

Application

A useful intermediate employed in the preparation of biologically active azoles and aminoguanidine derivatives.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?