• USA Home
  • 459410 - Polystyrene solution

EMAIL THIS PAGE TO A FRIEND
459410 Aldrich

Polystyrene solution

10 wt % dispersion in H2O, 3.0 μm average diameter, latex particles

Purchase

Properties

form   latex particles
contains   surface-bound sulfate groups as stabilizer
concentration   10 wt % dispersion in H2O
average diameter   3.0 μm

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?