EMAIL THIS PAGE TO A FRIEND
46088 Aldrich

Ethylsilane

≥97.0%

  • CAS Number 2814-79-1

  • Linear Formula CH3CH2SiH3

  • Molecular Weight 60.17

  •  Beilstein Registry Number 1730749

  •  MDL number MFCD00053748

  •  PubChem Substance ID 57650694

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organosilicon, Other Organosilicon
InChI Key   KCWYOFZQRFCIIE-UHFFFAOYSA-N
assay   ≥97.0%
bp   19 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1954 2.1
WGK Germany 
3
Flash Point(F) 
-76 °F
Flash Point(C) 
-60 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?