• USA Home
  • 461571 - (R)-(−)-1-Phenylethyl 3,5-dinitrobenzoate

EMAIL THIS PAGE TO A FRIEND
461571 Aldrich

(R)-(−)-1-Phenylethyl 3,5-dinitrobenzoate

98%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Esters, Organic Building Blocks More...
InChI Key   YATKDADIXNBBCZ-SNVBAGLBSA-N
assay   98%
optical activity   [α]22/D −47°, c = 2 in acetone
mp   122-125 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?