• USA Home
  • 461628 - (S)-(+)-N-[1-(1-Naphthyl)ethyl]-3,5-dinitrobenzamide

EMAIL THIS PAGE TO A FRIEND
461628 Aldrich

(S)-(+)-N-[1-(1-Naphthyl)ethyl]-3,5-dinitrobenzamide

97%

Purchase

Properties

Related Categories Amides, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents,
InChI Key   RUUPGHVIODHXRI-LBPRGKRZSA-N
assay   97%
optical activity   [α]22/D +112°, c = 2 in acetone
mp   224-226 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?