• USA Home
  • 461644 - (S)-(−)-N-[1-(1-Naphthyl)ethyl]succinamic acid

EMAIL THIS PAGE TO A FRIEND
461644 Aldrich

(S)-(−)-N-[1-(1-Naphthyl)ethyl]succinamic acid

98%

Purchase

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acids, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents,
InChI Key   AUSXHLAOQYCBAR-NSHDSACASA-N
assay   98%
optical activity   [α]24/D −45°, c = 2 in ethanol
mp   157-159 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?