• USA Home
  • 461679 - N,N-Bis[(R)-(+)-1-phenylethyl]phthalamic acid

EMAIL THIS PAGE TO A FRIEND
461679 Aldrich

N,N-Bis[(R)-(+)-1-phenylethyl]phthalamic acid

98%

Purchase

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acids, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents,
InChI Key   NCYIVOVUPRJYNJ-QZTJIDSGSA-N
assay   98%
optical activity   [α]22/D +153°, c = 1 in chloroform
mp   110 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?