EMAIL THIS PAGE TO A FRIEND
462802 Aldrich

2-Propyl-1H-indene

97%

Purchase

Properties

Related Categories Arenes, Building Blocks, Chemical Synthesis, Organic Building Blocks
InChI Key   DOUIKRLGKLTCJI-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.551(lit.)
bp   78 °C/0.5 mmHg(lit.)
density   0.95 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
219.2 °F
Flash Point(C) 
104 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?