CAS Number 20396-66-1
Linear Formula B2D6
Molecular Weight 33.71
MDL number MFCD00799509
PubChem Substance ID 24869898
| grade | electronic grade |
| assay | 9-11% (balance is deuterium) |
| concentration | 10% in D2 |
| bp | −92.5 °C(lit.) |
| mass shift | M+6 |
Diborane gas is used primarily as a p-type dopant in the deposition of epitaxial and amorphous silicon thin films. The unique properties of diborane-d6 can result in increased device performance and reliability.
48 L in steel cylinder
Stainless steel regulators Z527416 or Z527424 are recommended.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?