• USA Home
  • 463337 - Ethylene glycol methacrylate phosphate

EMAIL THIS PAGE TO A FRIEND
463337 Aldrich

Ethylene glycol methacrylate phosphate

Synonym: 2-(Methacryloyloxy)ethyl phosphate

Purchase

Properties

Related Categories Acrylic Monomers, Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis,
InChI Key   SEILKFZTLVMHRR-UHFFFAOYSA-N
contains   1000 ppm monomethyl ether hydroquinone as inhibitor
refractive index   n20/D 1.469(lit.)
density   1.31 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2920 3(8) / PGII
WGK Germany 
3
Flash Point(F) 
87.8 °F
Flash Point(C) 
31 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?