• USA Home
  • 464201 - Zirconium(IV) oxide-yttria stabilized

EMAIL THIS PAGE TO A FRIEND
464201 Aldrich

Zirconium(IV) oxide-yttria stabilized Green Alternative

submicron powder, 99.5% trace metals basis (purity excludes ~2% HfO2)

Synonym: YSZ, Yttria stabilized zirconia, Yttrium stabilized zirconium oxide

Purchase

Properties

Related Categories Alternative Energy, Chemical Synthesis, Electrolyte Materials, Electrolytes, Enabling Products,
InChI Key   KSTOPEBVPBSOTK-UHFFFAOYSA-N
assay   99.5% trace metals basis (purity excludes ~2% HfO2)
form   submicron powder
contains   ~5.3 mol % yttria as stabilizer
composition   (ZrO2)0.947(Y2O3)0.053
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
particle size   ~500 nm
surface area   6.9 m2/g
mp   >2,600 °C(lit.)

Description

Application

Electrolyte material for solid oxide fuel cells (SOFC).

Packaging

Packaged in glass bottles

Protocols & Applications

Advances in Materials for Solid Oxide Fuel Cells

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Advances in Materials for Solid Oxide Fuel Cells

As with all types of fuel cells, a Solid Oxide Fuel Cell (SOFC) is capable of efficiently transforming chemical energy into electrical energy. However, unlike fuel cells based on proton-conducting el...
Raymond J. Gorte
Material Matters Volume 5 Article 4
Keywords: Applications, Catalysis, Electrochemical reactions, Endothermic, Formulations, Gene expression, Methods, Oxidations, Support

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?