• USA Home
  • 464392 - (R)-(+)-4-(2-Chlorophenyl)-2-hydroxy-5,5-dimethyl-1,3,2-dioxaphosphorinane 2-oxide

EMAIL THIS PAGE TO A FRIEND
464392 Aldrich

(R)-(+)-4-(2-Chlorophenyl)-2-hydroxy-5,5-dimethyl-1,3,2-dioxaphosphorinane 2-oxide

97%

  • CAS Number 98674-87-4

  • Empirical Formula (Hill Notation) C11H14ClO4P

  • Molecular Weight 276.65

  •  MDL number MFCD00069409

  •  PubChem Substance ID 24870175

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Organic Phosphates/Phosphites More...
InChI Key   ZPSPULCZMWMHCY-JTQLQIEISA-N
assay   97%
optical activity   [α]22/D +46°, c = 1 in methanol
mp   228-234 °C(lit.)
storage temp.   2-8°C

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?