• USA Home
  • 464414 - (−)-Ouabain hydrate

EMAIL THIS PAGE TO A FRIEND

464414 Aldrich

(−)-Ouabain hydrate

98%

Synonym: G-Strophanthin

  • CAS Number 340169-01-9

  • Empirical Formula (Hill Notation) C29H44O12 · xH2O

  • Molecular Weight 584.65 (anhydrous basis)

  •  MDL number MFCD00149240

Purchase

Properties

assay   98%
optical activity   [α]20/D −25.0°, c = 1 in H2O
mp   185-192 °C(lit.)

Price and Availability

Customers Also Viewed

Digoxin
D6003Digoxin

analytical standard

Ouabain
O0200000Ouabain

European Pharmacopoeia (EP) Reference Standard

Ouabain octahydrate

≥95% (HPLC), powder

Safety & Documentation

Safety Information

Hazard Codes (Europe) 
T
Risk Statements (Europe) 
Safety Statements (Europe) 
45
RIDADR 
UN 1544 6.1/PG 1
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

FT-IR 1 (2), 1064:A / FT-NMR 1 (3), 607:B

Related Products

replaced by

Product #

Description

Add to Cart

O3125 Ouabain octahydrate, ≥95% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?