• USA Home
  • 464740 - Sodium dicyanoaurate(I)

EMAIL THIS PAGE TO A FRIEND
464740 Aldrich

Sodium dicyanoaurate(I)

97%

Synonym: Sodium gold cyanide

Purchase

Properties

Related Categories Essential Chemicals, Materials Science, Metal and Ceramic Science, Research Essentials, Salts,
InChI Key   LGYSFIGTWNEYKS-UHFFFAOYSA-N
assay   97%
form   powder

Description

Packaging

1 g in glass bottle

Price and Availability


Ultra-High Purity Inorganics
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
RIDADR 
UN 1588 6.1 / PGI
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?