• USA Home
  • 465429 - 2-(5-Isoxazolyl)phenol

EMAIL THIS PAGE TO A FRIEND
465429 Aldrich

2-(5-Isoxazolyl)phenol

98%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Isoxazoles
InChI Key   DBDXTIAEVFSDNN-UHFFFAOYSA-N
assay   98%
mp   183-186 °C(lit.)

Description

General description

2-(5-Isoxazolyl)phenol is an isoxazole derivative.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?