• USA Home
  • 465437 - 4-Chloro-2-(5-isoxazolyl)phenol

EMAIL THIS PAGE TO A FRIEND
465437 Aldrich

4-Chloro-2-(5-isoxazolyl)phenol

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks, Isoxazoles More...
InChI Key   AAJXJLYUWKMQNO-UHFFFAOYSA-N
assay   97%
mp   196-200 °C(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?