• USA Home
  • 465542 - Disodium [μ-(1,2-ethanediolato-O:O′)]tetrakis(1,2-ethanediolato-O,O′)disilicate

EMAIL THIS PAGE TO A FRIEND
465542 Aldrich

Disodium [μ-(1,2-ethanediolato-O:O′)]tetrakis(1,2-ethanediolato-O,O′)disilicate

97%

Synonym: Sodium glycolatosilicate

  • CAS Number 137704-78-0

  • Empirical Formula (Hill Notation) C10H22Na2O10Si2

  • Molecular Weight 404.43

  •  MDL number MFCD01632669

  •  PubChem Substance ID 24870286

Purchase

Properties

Related Categories Materials Science, Micro/NanoElectronics, Silicates, Silicon, Solution Deposition Precursors More...
InChI Key   RMWFMTQLKAETMI-UHFFFAOYSA-N
assay   97%
mp   >350 °C(lit.)

Description

Packaging

10, 50 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Bio-Inspired “Green” Synthesis of Nanomaterials and their Applications

Dr. Siddharth V. Patwardhan Department of Chemical and Process Engineering University of Strathclyde, 75 Montrose Street Glasgow G1 1XJ, UK Email: Siddharth.Patwardhan@strath.ac.uk
Keywords: Adsorption, Agriculture, Apoptosis, Cancer, Catalysis, Condensations, Cosmetics, Deposition, Environmental, Enzyme activity, Green chemistry, Hydration reaction, Immobilization, Microwave synthesis, Nanomaterials, Pharmaceutical, Precipitation, Reductions, Separation, Sol-gel, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?