• USA Home
  • 468487 - 3-Acetylnoradamantane

EMAIL THIS PAGE TO A FRIEND
468487 Aldrich

3-Acetylnoradamantane

technical grade, 85%

Purchase

Properties

Related Categories A, Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
grade   technical grade
InChI Key   VMCFOTARMDNPPQ-DTIDVZRVSA-N
assay   85%
refractive index   n20/D 1.502(lit.)
bp   212 °C(lit.)
density   1.044 g/mL at 25 °C(lit.)

Description

Packaging

500 mg in glass bottle

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
213.8 °F
Flash Point(C) 
101 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?