• USA Home
  • 469092 - DAB-Am-64, Polypropylenimine tetrahexacontaamine Dendrimer, Generation 5.0

EMAIL THIS PAGE TO A FRIEND
469092 Aldrich

DAB-Am-64, Polypropylenimine tetrahexacontaamine Dendrimer, Generation 5.0

volume 10,960 Å3

  • CAS Number 163611-05-0

  • Linear Formula [-CH2CH2N[(CH2)3N[(CH2)3N[(CH2)3N[(CH2)3N[(CH2)3NH2]2]2]2]2]2]2

  •  MDL number MFCD00803093

Purchase

Properties

Related Categories Dendrimers, Materials Science, Nanomaterials, Polypropylenimine Dendrimers
description   aminopropyl surface groups
mol wt   calculated mol wt 7,168
  generation 5
feature   core type 1,4-diaminobutane core (4-carbon core)
no. Surface Groups   64
volume   10,960 Å3
refractive index   n20/D 1.518(lit.)
viscosity   6.7 poise(50 °C)(lit.)
transition temp   Tg −84 °C
solubility   methanol and water: soluble
density   1.01 g/mL at 25 °C(lit.)

Description

Legal Information

DSM product

Physical form

Contains 64 primary amino groups.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2735PSN2 8 / PGIII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?