• USA Home
  • 469254 - α-Methyl-N-phenyl-1H-benzotriazole-1-methanamine

EMAIL THIS PAGE TO A FRIEND
469254 Aldrich

α-Methyl-N-phenyl-1H-benzotriazole-1-methanamine

97%

Purchase

Properties

Related Categories C-C Bond Formation, Chemical Synthesis, Other Synthetic Reagents for C-C Bond Formation, Synthetic Reagents
InChI Key   HLIOCSMHAVCKMV-UHFFFAOYSA-N
assay   97%
mp   128-131 °C(lit.)

Description

Application

Intermediate in the preparation of secondary aromatic amines, α-aminoalkylketones, and substituted tetrahydroquinolines.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Indoles, Purines, and Their Isosteres

Substituted indoles and purines have frequently been referred to as “privileged structures” since they are capable of binding to multiple receptors with high affinity, and thus have applications acro...
Keywords: Building blocks, Medicinal chemistry

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?