• USA Home
  • 470155 - 2-(Difluoromethoxy)benzaldehyde

EMAIL THIS PAGE TO A FRIEND
470155 Aldrich

2-(Difluoromethoxy)benzaldehyde

96%

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C8, Carbonyl Compounds, Chemical Synthesis,
InChI Key   QPBNHDFPMRENBC-UHFFFAOYSA-N
assay   96%
refractive index   n20/D 1.497(lit.)
bp   216 °C(lit.)
density   1.3 g/mL at 25 °C(lit.)

Description

Packaging

5 g in glass bottle

Application

2-(Difluoromethoxy)benzaldehyde may be used as a starting reagent in the synthesis of the following compounds:
• 2-[2-(difluoromethoxy)benzylidene]methyl acetoacetate
• 3-(2-cyanoethyl) 5-(2-propoxyethyl) 4-[2-(difluoromethoxy)phenyl]-2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate
• 2-propoxyethyl 5-cyano-4-[2-(difluoromethoxy)phenyl]-2-methyl-6-thioxo-1,4,5,6-tetrahydropyridine-3-carboxylate
• methyl 5-cyano-4-(2-difluoromethoxyphenyl)-2-methyl-6-thioxo-1,4,5,6-tetrahydropyridine-3-carboxylate

General description

2-(Difluoromethoxy)benzaldehyde is an ortho-difluoromethoxy substituted benzaldehyde.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
224.6 °F
Flash Point(C) 
107 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?