• USA Home
  • 470473 - tert-Dodecyl polysulfide

EMAIL THIS PAGE TO A FRIEND
470473 Aldrich

tert-Dodecyl polysulfide

average Mn ~498, average molecular diameter 10.76 Å, TPS® 32

Synonym: Di-tert-dodecyl polysulfide, TPS® 20

Purchase

Properties

Related Categories Materials Science, Polymer Additives, Polymer Property Modifiers, Polymer Science
mol wt   average Mn ~498
mfr. no.   TPS® 32
refractive index   n20/D 1.545
pore size   10.76 Å average molecular diameter
viscosity   120 cP(40 °C)(lit.)
transition temp   pour point −9 °C
density   1.01 g/mL at 25 °C

Description

Legal Information

Product of Arkema Inc.

TPS is a registered trademark of Arkema France

Price and Availability

Safety & Documentation

Safety Information

Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
291.2 °F
Flash Point(C) 
144 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

470465 tert-Dodecyl polysulfide, average Mn ~434, TPS 20

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?