• USA Home
  • 47355 - 4-Fluoro-D-α-phenylglycine

EMAIL THIS PAGE TO A FRIEND
47355 Aldrich

4-Fluoro-D-α-phenylglycine

≥99.0% (NT)

  • CAS Number 93939-74-3

  • Empirical Formula (Hill Notation) C8H8FNO2

  • Molecular Weight 169.15

  •  Beilstein Registry Number 5850632

  •  EC Number 300-363-8

  •  MDL number MFCD00042727

  •  PubChem Substance ID 57650931

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylglycine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   JKFYKCYQEWQPTM-SSDOTTSWSA-N
assay   ≥99.0% (NT)
optical activity   [α]20/D −138±2°, c = 1% in 1 M HCl
mp   ≥300 °C

Description

Packaging

1 g in glass bottle

100 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?