• USA Home
  • 474029 - Poly(methyl methacrylate-co-butyl methacrylate)

EMAIL THIS PAGE TO A FRIEND
474029 Aldrich

Poly(methyl methacrylate-co-butyl methacrylate)

methyl methacrylate ~85 mol %, average Mw ~75,000

Purchase

Properties

Related Categories Acrylics, Bioelectronics, Flexible Organic Transistors, Hydrophobic Polymers, Materials Science,
mol wt   average Mw ~75,000
composition   methyl methacrylate, ~85 mol %
reduced viscosity   35-40 cm3/g(20 °C)(lit.)
acid number   <1 mg KOH/g
transition temp   Tg (DSC) 105 °C (midpoint)
solubility   alcohols and aliphatic hydrocarbons: insoluble
  esters, ketones, aromatics and glycol ethers: soluble
density   1.15 g/mL at 25 °C(lit.)

Description

General description

∼85 mole% MMA

Application

Pigment-wetting binder for coatings on plastics.

Features and Benefits

Contributes hardness, weather resistance, light fastness and resistance to chemicals.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?