• USA Home
  • 47402 - Fmoc-Gln(Trt)-Wang resin

EMAIL THIS PAGE TO A FRIEND
47402 Aldrich

Fmoc-Gln(Trt)-Wang resin

extent of labeling: 0.4-0.8 mmol/g loading

Synonym: Nα-Fmoc-Nδ-trityl-L-glutamine 4-benzyloxybenzyl ester polymer-bound

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Polymer Supports for SPPS, Pre-Loaded Resins,
extent of labeling   0.4-0.8 mmol/g loading
matrix   polystyrene, crosslinked with 1% DVB
particle size   100-200 mesh
storage temp.   2-8°C

Description

Packaging

1 g in poly tube

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Wang resins Preloaded with N-α-Fmoc-Protected Amino Acids B

TFA-labile Wang resins are standard supports for batch synthesis of peptide acids following the Fmoc-/tBu-protection scheme.(1,2) The Fmoc-amino acids are coupled to the 4-hydroxymethylphenoxyacetic ...
Chemfiles Volume 3 Article 4
Keywords: Epimerizations, Methods, Peptide synthesis, Protocols, Solid phase peptide synthesis

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?