• USA Home
  • 474134 - [2,2,3,3,4,4,5,5,6,6,7,7,8,9,9,9-Hexadecafluoro-8-(trifluoromethyl)nonyl]oxirane

EMAIL THIS PAGE TO A FRIEND
474134 Aldrich

[2,2,3,3,4,4,5,5,6,6,7,7,8,9,9,9-Hexadecafluoro-8-(trifluoromethyl)nonyl]oxirane

96%

Purchase

Properties

Related Categories Epoxide Monomers, Fluorine-Containing Monomers for 157 nm UV Lithography Resist Polymers, Lithography Monomers, Materials Science, Micro/NanoElectronics,
InChI Key   ZFJPGQQBBNLQOX-UHFFFAOYSA-N
assay   96%
refractive index   n20/D 1.324(lit.)
bp   93-94 °C/8 mmHg(lit.)
density   1.757 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?