• USA Home
  • 474282 - 4,4,5,5,6,6,7,7,8,9,9,9-Dodecafluoro-2-hydroxy-8-(trifluoromethyl)nonyl methacrylate

EMAIL THIS PAGE TO A FRIEND
474282 Aldrich

4,4,5,5,6,6,7,7,8,9,9,9-Dodecafluoro-2-hydroxy-8-(trifluoromethyl)nonyl methacrylate

97%

  • CAS Number 16083-81-1

  • Linear Formula H2C=C(CH3)CO2CH2CH(OH)CH2(CF2)4CF(CF3)2

  • Molecular Weight 512.21

  •  MDL number MFCD00236099

  •  PubChem Substance ID 24870981

Purchase

Properties

Related Categories Acrylic Monomers, Fluorinated Acrylics, Fluorine-Containing Monomers for 157 nm UV Lithography Resist Polymers, Lithography Monomers, Materials Science,
InChI Key   RPDBRTLKDYJCCE-UHFFFAOYSA-N
assay   97%
contains   100 ppm 4-tert-butylcatechol as inhibitor
refractive index   n20/D 1.368(lit.)
bp   270 °C(lit.)
density   1.561 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?