• USA Home
  • 474568 - Poly(4-vinylphenol), brominated

EMAIL THIS PAGE TO A FRIEND
474568 Aldrich

Poly(4-vinylphenol), brominated

average Mn ~2,800, average Mw ~5,800

Purchase

Properties

Related Categories Bioelectronics, Flexible Organic Transistors, Hydrophobic Polymers, Materials Science, Poly(4-vinylphenol) (PVP),
mol wt   average Mn ~2,800
  average Mw ~5,800
extent of labeling   ~50 wt. % Br
transition temp   Tg (DSC) 120 °C (onset)
density   1.9 g/mL at 25 °C(lit.)

Description

Application

Fire retardant plastics additive and epoxy hardener. Photosensitive resin for printing plates and photoresists.

Price and Availability


Webinar: Recent Developments in RAFT Agents
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?