EMAIL THIS PAGE TO A FRIEND
47535 Aldrich

Fmoc-His(Tos)-OH

purum, ≥98.0% (calc. based on dry substance, C/N)

Synonym: Nα-Fmoc-N(im)-tosyl-L-histidine

  • CAS Number 112380-10-6

  • Empirical Formula (Hill Notation) C28H25N3O6S

  • Molecular Weight 531.58

  •  Beilstein Registry Number 5195552

  •  MDL number MFCD00144353

  •  PubChem Substance ID 24871115

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Histidine, Peptide Chemistry More...
InChI Key   PHAKSHNECOECCD-SANMLTNESA-N
grade   purum
assay   ≥98.0% (calc. based on dry substance, C/N)
optical activity   [α]20/D −7±1°, c = 1% in DMF
mp   170-174 °C (dec.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?