• USA Home
  • 475491 - 6-Acetyl-2H-1,4-benzoxazin-3(4H)-one

EMAIL THIS PAGE TO A FRIEND
475491 Aldrich

6-Acetyl-2H-1,4-benzoxazin-3(4H)-one

98%

Purchase

Properties

Related Categories Benzoxazines, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
InChI Key   BKJFWHFUERNXLJ-UHFFFAOYSA-N
assay   98%
mp   193-195 °C(lit.)

Description

Packaging

5 g in glass bottle

General description

6-Acetyl-2H-1,4-benzoxazin-3(4H)-one, also referred to as 6-acetyl-2H-benzo[b][1,4]oxazin-3(4H)-one, is a useful organic building block.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?