• USA Home
  • 475718 - 1,2-Propanediol diglycidyl ether

EMAIL THIS PAGE TO A FRIEND
475718 Aldrich

1,2-Propanediol diglycidyl ether

  • CAS Number 16096-30-3

  • Empirical Formula (Hill Notation) C9H16O4

  • Molecular Weight 188.22

  •  MDL number MFCD01861203

Purchase

Properties

InChI Key   HDPLHDGYGLENEI-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?